
Tungsten ≥99% (metals basis), powder -50+100 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-33-7
:
Formula:W
MDL Number:MFCD00011461
:183.84 g/mol
Storage Temperature:Ambient
EINECS:231-143-9
UN:3089
ADR:4.1,III
:GHS02
Frequently Bought Together