ContactUsLinkComponent
Quantus® Protect Xtra, Cleanroom Coats with Zip Closure
Quantus® Protect Xtra, Cleanroom Coats with Zip Closure
Supplier:  VWR Collection
Certificates
undefined
Quantus® Protect Xtra, Cleanroom Coats with Zip Closure
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 113-9957
  • 113-9955
  • 113-9956
  • 113-9953
  • 113-9952
  • 113-9954
  • 113-9958
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Fabric:
  • Protect Xtra fabric (100% polyester weave with conductive yarn (warp))
  • Protect Xtra fabric (100% polyester weave with conductive yarn (warp))
  • Protect Xtra fabric (100% polyester weave with conductive yarn (warp))
  • Protect Xtra fabric (100% polyester weave with conductive yarn (warp))
  • Protect Xtra fabric (100% polyester weave with conductive yarn (warp))
  • Protect Xtra fabric (100% polyester weave with conductive yarn (warp))
  • Protect Xtra fabric (100% polyester weave with conductive yarn (warp))
  • Weight:
  • 98,3 g/m²
  • 98,3 g/m²
  • 98,3 g/m²
  • 98,3 g/m²
  • 98,3 g/m²
  • 98,3 g/m²
  • 98,3 g/m²