Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solution, pH 6,865

Buffer solution, 6,865, ±0,01, 25 °C, 1 L
Catalog # APPC277124.1211
Supplier:   PanReac AppliChem
Buffer solution, pH 6,865
Catalog # APPC277124.1211
Supplier:  PanReac AppliChem
CAS Number:  
Buffer solution, pH 6,865
Buffer solution, pH 6,865

Buffer solution, 6,865, ±0,01, 25 °C, 1 L

CAS Number:  
Price
52,80 €
Log in for availability
1 * 1 L
Product Details & Documents

These buffer solutions are useful for the calibration of high precision laboratory pH meters.

  • pH (30 °C): 6,853 ± 0,01
  • pH (25 °C): 6,865 ± 0,01
  • pH (20 °C): 6,881 ± 0,01
  • pH (10 °C): 6,900 ± 0,01

 : Standardised, according to DIN19266.

Specifications

Specifications

  • Pk:
    1 L
  • Reference temperature:
    25 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,01
  • pH value:
    6,865

Specifications