Order Entry
Belgium
ContactUsLinkComponent
Magnesium ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Magnesium ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40859.KS
CAS Number:  
Magnesium ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40859.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7439-95-4
 :
Formula:Mg
MDL Number:MFCD00085308
 :24.3 g/mol
Merck Index:13,05675
Boiling Pt:1107 °C (1013 hPa)
Melting Pt:651 °C
Density:1.74 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-104-6
UN:1869
ADR:4.1,III