
Formamide, OmniPur®, Deionized, Millipore®
Catalog # CALB4610-100ML
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:45.04 g/mol
:GHS08
Storage Temperature:Refrigerator
CAS Number:75-12-7
Formula:HCONH₂
MDL Number:MFCD00007941