Orders LinkContactUsLinkComponent
Sample vials
Sample vials
Sample vials, 12,00 ml, No
Catalog # PAAR21550
Supplier:   Anton Paar
Sample vials
Catalog # PAAR21550
Supplier:  Anton Paar
Sample vials
Sample vials
Sample vials, 12,00 ml, No
Price
686,00 €
Log in for availability
1 * 2.000 ST
Product Details & Documents
These sample vials made of plastic are used with a sample changer (1/48/96 positions magazine) to automatically fill sample into the measuring instrument.
Specifications
  • Cap included:
    No
  • Capacity:
    12,00 ml
  • Description:
    Sample vials