Order Entry
Belgium
ContactUsLinkComponent
pH electrode calibration and maintenance solutions kit
pH electrode calibration and maintenance solutions kit
Catalog # 662-0322
Supplier:  WTW
CAS Number:  
undefined
pH electrode calibration and maintenance solutions kit
Catalog # 662-0322
Supplier:  WTW
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 SET
  • Description:
    Buffers, electrolyte and cleaning solutions (3×250 ml)
  • Seq:
    01
  • pH value:
    4,006/6,865/9,180
  • Contents:
    1 bottle with 250 ml pepsin cleaning solution, 1 bottle with 250 ml KCl solution 3 mol/L

Specifications

About this item

Adapted to different electrode types for calibration and maintenance.

The pepsin cleaning solution is used to remove protein containing precipitation from diaphragms, the 3 molar KCl as a storage or refill solution.

Delivery: Kit contains set of 250 ml each of pepsin cleaning solution; 3 M KCl solution and standard buffer solutions of pH 4,006, 6,865, 9,180.