Orders LinkContactUsLinkComponent
Redox buffer solutions
Redox buffer solutions
Redox buffer solutions
Redox buffer solutions are used for verification of all common redox sensors.
  • Temperature dependent
  • Used in quality assurance systems
A table on every bottle label indicates redox values at different temperatures and hence not used for calibration purposes.
 : Supplied with a quality inspection certificate.
Product Family Options
  • Description
    pH value
    Pk Info
    Pk
  • Redox buffer, 468 mV
    0,1
    6×30 ml
    6
    1 * 6 ST
    1 * 6 ST - 38,20 €
    38,20 €
    1 * 6 ST
       38,20 €
  • Redox buffer, 220 mV
    7
    1×250 ml
    250 mL
    1 * 250 mL
    1 * 250 mL - 40,60 €
    40,60 €
    1 * 250 mL
       40,60 €
  • Redox buffer, 220 mV
    7
    6×250 ml
    6
    1 * 6 ST
    1 * 6 ST - 178,00 €
    178,00 €
    1 * 6 ST
       178,00 €