Soil test kit, NPK
Catalog # LAMO3-5880
Supplier: LaMotte Chemicals
CAS Number:
Specifications
- Pk:1
- Description:NPK soil text kit
- Environmentally Preferable:
Specifications
About this item
Kit is sufficient to perform up to fifty tests each for nitrogen, phosphorus, and potassium levels using non-hazardous TesTabs.
- Kit is fun to use and easy to ship anywhere
- Ideal for upper elementary, middle, and high school students
It is specifically designed to meet the needs of the GLOBE ProgramTM (Global Learning and Observations to Benefit the Environment).
Delivery: It includes the necessary apparatus, simple diagrammed instructions, and a laminated colour chart.