Orders LinkContactUsLinkComponent
Precision thermometers
Precision thermometers
Supplier:   Amarell
Precision thermometers
Supplier:  Amarell
Certificates
Precision thermometers
2 Options Available Below
Product Details & Documents
These thermometers are suitable for government verification.
  • Round top finish in transparent square plastic case
  • Durable pigment
  • Solid stem and total immersion
Certifications: Supplied with DIN 12785 certificate.
Documents
Product Family Options
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • AMARM247302-FLDKD3
    Precision thermometer, white, with DKD3 certificate
    Precision thermometer, white, with DKD3 certificate
    +50...+150 °C
    ±0,5 °C
    300 mm
    Red
    6,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 318,00 €
    318,00 €
    1 * 1 ST
       318,00 €
  • AMARM247302-FL
    Precision thermometer, white
    Precision thermometer, white
    +50...+150 °C
    ±0,5 °C
    300 mm
    Red
    6,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 93,30 €
    93,30 €
    1 * 1 ST
       93,30 €