Orders LinkContactUsLinkComponent
Cryo apron
Blue, One size
Catalog # AIRLACCSECU19
Supplier:   Air Liquide
Cryo apron
Catalog # AIRLACCSECU19
Supplier:  Air Liquide
Cryo apron
Blue, One size
Price
320,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This apron with neck tie is ideal for use in environments where cryogenic fluids are a high risk factor for cryogenic burns.
Specifications
  • Clothing Size:
    One size
  • Colour:
    Blue
  • Cat. No.:
    AIRLACCSECU19
  • Risk area:
    Cryo
  • Garment application:
    Laboratory|Industrial
  • Gender:
    Unisex