Slide stainer
Slide stainer and dyer, SS-30, 30
Catalog # KLINSS-30
Supplier: KLINIPATH
Jump to:
Product Details & Documents
Slide stainer designed to be used for Hematoxylin and Eosin (H&E), Papanicolau (PAP) and user-specified staining protocols.
- Bilateral robotic arm: Free choice of the station sequence in a staining protocol
- Staining capacity of up to five racks simultaneously, depending on protocols, load frequency and instrument configuration
- USB connection for programming of staining protocols and data gathering
The Reagent Management System (RMS) controls the use of reagents and helps to achieve a better staining quality.
Documents
Myr SLIDE STAINER SS 30
Videos
Specifications
- Model:SS-30
- No. of slides:30
- Description:Slide stainer and dyer