Drigalski spatula
172 mm, Green
Supplier: NEOLAB
Jump to:
Product Details & Documents
This disposable Drigalski spatula made from ABS features a triangular head (14 mm).
- Smooth surface does not destroy agar and does not mix media layers
- Triangular head distributes the pressure evenly
- Handle is slightly flexible and equipped with grip grooves
Packaging: Individually sterile packed.
Documents
Certificates
Search by Lot NumberSpecifications
- Colour:Green
- Length:172 mm



