Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cooler, Powerbox® Plus

Cooler, Powerbox® Plus

Electric Cooler, Weight: 3,6 kg, 24 l, 410×320×420 mm
Catalog # CAMP2000024955
Supplier:   CAMPING GAZ
Cooler, Powerbox® Plus
Catalog # CAMP2000024955
Supplier:  CAMPING GAZ
Cooler, Powerbox® Plus

Electric Cooler, Weight: 3,6 kg, 24 l, 410×320×420 mm

Price
118,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The Powerbox® Plus electric cooler is packed with 30% more premium insulation than its predecessor and keep content 20 °C below the ambient temperature and the fan emits a maximum of 39 dB.

  • Thicker and improved PU insulation
  • Firmly locking lid
  • Noise reduced fan

Cooler Powerbox® Plus has antimicrobial lining that comes from a chemical additive injected into the plastic within the liner.

Specifications
  • Capacity:
    24 l
  • Description:
    Electric Cooler, Weight: 3,6 kg
  • Dimensions:
    410×320×420 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.