Silicone antifoam A
:
Frequently Bought Together
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
: MERCK PRODUCTION CHEMICALS
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Toluene ≥98%, TECHNICAL
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
: VWR Chemicals
Glycerine (synthetic) 99+%, pure
: Thermo Fisher Scientific
Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
N-Bromosuccinimide 99%
: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9+%, AcroSeal®
: Thermo Fisher Scientific