pH electrodes, Sentix® IDS
: WTW
High quality, enhanced sensor technology combined with most modern measurement electronics.
- Intelligent: Electrodes are automatically recognized by the meter, each sensor is exactly identified
- Digital: Measurement signals are processed digitally, providing very accurate data
- Measurement integrity benchmarking of the electrodes by QSC (Quality Sensor Control)
- IDS electrodes store their serial number and their calibration records
Ø×L: 12×120 mm
: Various cable lengths available.
High quality, enhanced sensor technology combined with most modern measurement electronics.
- Intelligent: Electrodes are automatically recognized by the meter, each sensor is exactly identified
- Digital: Measurement signals are processed digitally, providing very accurate data
- Measurement integrity benchmarking of the electrodes by QSC (Quality Sensor Control)
- IDS electrodes store their serial number and their calibration records
Ø×L: 12×120 mm
: Various cable lengths available.
Product Family Options
- DescriptionTypepHTemp.MaterialElectrolyteDiaphr.Ø×LConnection
- IDS pH electrode Sentix® 940Sentix® 9400,000 - 14,000 (±0,004)0...+80 °CPlasticGelFibre12×120 mm1,5 m cable1 * 1 ST333,00 €
- IDS pH electrode Sentix® 940-3Sentix® 940-30,000 - 14,000 (±0,004)0...+80 °CPlasticGelFibre12×120 mm3 m cable1 * 1 ST376,00 €
- IDS pH electrode Sentix® 950Sentix® 9500,000 - 14,000 (±0,004)0...+80 °CPlastic3M KClCeramic12×120 mm1,5 m cable1 * 1 ST403,00 €
- IDS pH electrode Sentix® 980Sentix® 9800,000 - 14,000 (±0,004)0...+100 °CGlass3M KClPlatinum wire12×120 mm1,5 m cable1 * 1 ST376,00 €
Armouring
- Description
- Armoring A 925/K, with plastic protective hood, for TetraCon® 925-P, SensoLyt® 900-P, SensoLyt® ORP 900-P and FDO® 925-P1 * 1 ST361,00 €