Adrenergic ligand library, Screen-Well®
Supplier: Enzo Life Sciences
Certificates
About this item
The adrenergic ligand library contains 84 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein-coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The adrenergic ligand library is 1 plate of the larger neurotransmitter library which contains over 650 CNS receptor ligands.
Frequently Bought Together