Z-Pro-Nle-Asp-CHO
ENZOBMLZW94900100
New Product
:
- Pk:100 µG
- Source:
- Species:
- Seq:02
- Environmentally Preferable:
- Protein/peptide name:Z-Pro-Nle-Asp-CHO
Highly specific aldehyde based inhibitor for the caspase-like peptidic activity of the 20S proteasome.