PI 3K SH2 OMT
Catalog # ENZOBMLST4250500
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:0,5 mg
- Source:
- Species:
- Seq:02
- Environmentally Preferable:
- Protein/peptide name:PI 3K SH2 OMT
Specifications
About this item
PI3 kinase inhibitor
This unique SH2 domain ligand is derived from the sequence flanking Tyr751 of the PDGF receptor. It inhibits the PI 3-kinase p85 C-terminal SH2 domain (IC50=14,2 µM). It contains the nonhydrolyzable phosphotyrosine mimetic L-O-(2-malonyl)tyrosine (OMT) and is provided as the trimethyl ester for greater cell permeability.