Order Entry
Belgium
ContactUsLinkComponent
Human alpha2-macroglobulin (plasma) (from plasma)
Human alpha2-macroglobulin (plasma) (from plasma)
Catalog # ENZOBMLSE5020001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human alpha2-macroglobulin (plasma) (from plasma)
Catalog # ENZOBMLSE5020001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
    Plasma
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    alpha2-Macroglobulin (plasma)

Specifications

About this item

Protease inhibitor

Isolated from human plasma that is tested to be negative for HbsAg, anti-HBc, anti-HCV, and anti-HIV-1 and -2.