Order Entry
Belgium
ContactUsLinkComponent
Int-H1-S6A F8 c-myc inhibitor
Int-H1-S6A F8 c-myc inhibitor
Catalog # ENZOBMLP6050500
Supplier:  Enzo Life Sciences
CAS Number:  
Int-H1-S6A F8 c-myc inhibitor
Catalog # ENZOBMLP6050500
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    0,5 mg
  • Seq:
    02
  • Protein/peptide name:
    Int-H1-S6A F8 c-myc inhibitor
  • Formulation:
    Lyophilised solid

Specifications

About this item

c-Myc inhibitor

H1 DNA-binding region of c-Myc containing Ser to Ala, and Phe to Ala substitutions (underlined) to confer an increase in its potency to inhibit c-Myc. The N-terminus of this peptide is the Int peptide sequence derived from the third Antennapedia homeodomain, to confer cell permeability. Control for this peptide is H1-S6A,F8A c-Myc inhibibitor peptide (Prod. No. BML-P606). Inhibited cloning efficiency of MCF-7 human breast cancer cells by 90% at 10 µM (IC50=5,9 µM). In MCF-7 cells, it inhibited cell growth and induced apoptosis. In addition, at 10 µM it strongly inhibited transcription of the c-Myc regulated genes ODC and p53. The non-cell-permeable control for this peptide is H1-S6A, F8A c-Myc inhibitor peptide which lacks the N-terminal internalization sequence.