Ac-Ala-Pro-Nle-Asp-H
Catalog # ENZOBMLAW94850100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Seq:02
- Protein/peptide name:Ac-Ala-Pro-Nle-Asp-H
Specifications
About this item
Highly specific aldehyde based inhibitor of the caspase-like peptidic activity of the 20S proteasome.