ContactUsLinkComponent
MMP Inhibitor profiling kit, fluorometric
MMP Inhibitor profiling kit, fluorometric
Catalog # ENZOBMLAK0160001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
MMP Inhibitor profiling kit, fluorometric
Catalog # ENZOBMLAK0160001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    MMP Inhibitor profiling kit, fluorometric
  • Seq:
    02
  • Environmentally Preferable:
Specifications
About this item
A QUANTIZYME® Assay System Complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes. The assays are performed in a convenient 96-well microplate format using a sensitive fluorogenic substrate . Included are active MMP enzymes, assay buffer, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet.