ContactUsLinkComponent
MMP Inhibitor profiling kit, colorimetric
  ENZOBMLAK0150001
 :  
MMP Inhibitor profiling kit, colorimetric
  ENZOBMLAK0150001
 :  
  • Pk:
    1 KIT
  • Description:
    MMP Inhibitor profiling kit, colorimetric
  • Seq:
    02
A QUANTIZYME® Assay System Complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes. The assays are performed in a convenient 96-well microplate format using a chromogenic substrate1,2 detected at 412 nm. Included are active MMP enzymes, assay buffer, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet.