PKCζ PseudoSubstrate
ENZOALX260155M001
:
- Pk:1 mg
- Seq:02
- Protein/peptide name:PKCζ PseudoSubstrate
- Formulation:Counter Ion: TFA
Selective cell permeable inhibitor of PKCzeta. Activates eNOS (NOS III) through Akt phosphorylation and can be considered as a nitric oxide production activator in endothelial cells.
Selective cell permeable inhibitor of PKCzeta. Activates eNOS (NOS III) through Akt phosphorylation and can be considered as a nitric oxide production activator in endothelial cells.