Biotinyl-DEVD-CHO
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Inhibitor of caspase-3 and -7
Reversible inhibitor of caspase-3 and -7. Useful for the purification or extract depletion of caspase-3 in conjunction with streptavidin conjugates.
Specifications
- Protein/peptide name:Biotinyl-DEVD-CHO
- Purity:>99% (HPLC)
- Formulation:Lyophilized
Specifications
Frequently Bought Together