Brain (rat) tissue extract
Rat brain tissue extract
Catalog # ENZOADILYTRB100F
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
- Liquid
Delivery information: SDS-PAGE sample buffer (pH: 6,8) containing 40 mM TRIS-HCl, 1% SDS, 50mM DTT, 7,5% glycerol and 0,003% bromophenol blue.
Specifications
- Description:Rat brain tissue extract
- Cat. No.:ENZOADILYTRB100F



