Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Cholera Toxin (
Vibrio cholerae
) G Protein Activator
Cholera Toxin (Vibrio cholerae) G Protein Activator
ENZOBMLG1170001
:
ENZO LIFE SCIENCES
:
Cholera Toxin (Vibrio cholerae) G Protein Activator
ENZOBMLG1170001
:
ENZO LIFE SCIENCES
:
Pk:
1 mg
CAS No.:
9012-63-9
Seq:
02
Enzyme name:
Cholera Toxin (Vibrio cholerae) G protein activator
G protein activator.
Lyophilized.