Human recombinant Isopeptidase T (long form) (from E. coli)
Catalog # ENZOBMLUW96900025
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Enzyme name:Isopeptidase T (long form)
Specifications
About this item
Produced in E. coli.