Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Cu/Zn superoxide dismutase (from E. coli)
Human recombinant Cu/Zn superoxide dismutase (from E. coli)
Catalog # ENZOBMLSE3050100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant Cu/Zn superoxide dismutase (from E. coli)
Catalog # ENZOBMLSE3050100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Enzyme name:
    Cu/Zn superoxide dismutase

Specifications

About this item

Produced in E. coli.