Order Entry
Belgium
ContactUsLinkComponent
Human Neutrophil elastase (from neutrophils)
Human Neutrophil elastase (from neutrophils)
Catalog # ENZOBMLSE2840100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Human Neutrophil elastase (from neutrophils)
Catalog # ENZOBMLSE2840100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Source:
    Neutrophils
  • Species:
    Human
  • Seq:
    02
  • Enzyme name:
    Neutrophil elastase
  • Purity:
    >95% (SDS-PAGE). Although this is in highly purified form, and the source material tested negative for HbsAg, anti-HBc, anti-HCV, and anti-HIV-1 and -2, handle this product with all due precautions.

Specifications

About this item

Isolated from human neutrophils. Active enzyme.

  • Functions in inflammation

Can cleave a variety of proteins, including fibrous elastin, collagen, clotting factors, TNF-α , and α 1- antichymotrypsin. Because of its potential roles in inflammatory diseases such as emphysema and cystic fibrosis, it is a target for drug discovery.