Human recombinant Caspase-7 (from E. coli)
Catalog # ENZOBMLSE1775000
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 Vial
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Form:Liquid
- Enzyme name:Caspase-7
- Enzyme activity:5000 U
- Purity:>95% (SDS-PAGE)
- Concentration:100 U/µl
- Formulation:In 50 mM HEPES, pH 7,4, containing 100 mM sodium chloride, 0,5% CHAPS, 1 mM EDTA, 10% glycerol and 10 mM DTT.
Specifications
About this item
Produced in E. coli. Human caspase-7 (aa 24-303). Genbank Accession No. U40281.