Human recombinant Caspase-9 (from E. coli)
Catalog # ENZOBMLSE1735000
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 Vial
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Enzyme name:Caspase-9
- Enzyme activity:5000 U
Specifications
About this item
Produced in E. coli. C-terminal Human caspase-9 (aa 131-416).