Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Caspase-3 (from E. coli)
Human recombinant Caspase-3 (from E. coli)
Catalog # ENZOBMLSE1695000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant Caspase-3 (from E. coli)
Catalog # ENZOBMLSE1695000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Enzyme name:
    Caspase-3
  • Enzyme activity:
    5000 U
  • Purity:
    ≥95% (SDS-PAGE)
  • Formulation:
    100 U/µl in 50 mM HEPES, pH 7,4, 100 mM sodium chloride, 0,5% CHAPS, 1 mM EDTA, 10% glycerol and 10 mM dithiothreitol.

Specifications

About this item

Produced in E. coli. The enzyme is cleaved and activated from the proenzyme.

Caspase-3 (CPP32; Yama; apopain) from human cDNA, expressed in an E. coli expression system. Can be used to study enzyme regulation and kinetics, cleave target substrates and screen for inhibitors.