Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Caspase-1 (from E. coli)
Human recombinant Caspase-1 (from E. coli)
Catalog # ENZOBMLSE1685000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Human recombinant Caspase-1 (from E. coli)
Catalog # ENZOBMLSE1685000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Form:
    Liquid
  • Enzyme name:
    Caspase-1
  • Enzyme activity:
    5000 U
  • Purity:
    >90% (SDS-PAGE)
  • Concentration:
    100 U/μl
  • Formulation:
    In 50 mM HEPES, pH 7,4, containing 100 mM sodium chloride, 0,5% CHAPS, 1 mM EDTA, 10% Glycerol and 10 mM DTT.

Specifications

About this item

Produced in E. coli.

cDNA encodes residues identical to Asn120-His404 (C-terminus) at Genbank Accession No. M87507, except for an Asp381 to Glu change, introduced to stabilize the enzyme against autoproteolysis.