Staphylococcus aureus sphingomyelinase
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Isolated from Staphylococcus aureus.
Specifications
- Species:Staphylococcus aureus
- Form:Liquid
- Enzyme name:Sphingomyelinase
- Enzyme activity:100 - 300 U/mg
- Formulation:Supplied in 50% glycerol/0,25 M phosphate buffer
Specifications
Frequently Bought Together