Recombinant Caspase (active) set lll
Catalog # ENZOALX850243KI01
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 SET
- Enzyme type:Recombinant
- Seq:02
- Form:Lyophilized
- Reconstitution instructions:Reconstitute to 0,1 - 1 U/µl in PBS.
- Enzyme name:Caspase (active) set lll
- Purity:All components ≥90% (SDS-PAGE)
Specifications
About this item
We recommend using 1 U per assay for analyzing caspase activity.
Active recombinant caspases are useful in screening caspase inhibitors, studying enzyme kinetics and regulation, determining substrate specificity, and as a positive control in caspase activity assays.
Ordering information: Contents: Contains 25 units each of the following caspases:
- Caspase-1 (active) (human) (recombinant), yellow cap
- Caspase-2 (active) (human) (recombinant), orange cap
- Caspase-3 (active) (human) (recombinant), red cap
- Caspase-6 (active) (human) (recombinant), violet cap
- Caspase-7 (active) (human) (recombinant), red cap
- Caspase-8 (active) (human) (recombinant), green cap
- Caspase-9 (active) (human) (recombinant), blue cap
- Caspase-10 (active) (human) (recombinant), amber cap
- Caspase-1 (active) (human) (recombinant), yellow cap
- Caspase-2 (active) (human) (recombinant), orange cap
- Caspase-3 (active) (human) (recombinant), red cap
- Caspase-6 (active) (human) (recombinant), violet cap
- Caspase-7 (active) (human) (recombinant), red cap
- Caspase-8 (active) (human) (recombinant), green cap
- Caspase-9 (active) (human) (recombinant), blue cap
- Caspase-10 (active) (human) (recombinant), amber cap