Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Matriptase
Human recombinant Matriptase
Catalog # ENZOALX201246U250
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant Matriptase
Catalog # ENZOALX201246U250
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Enzyme type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Enzyme name:
    Matriptase
  • Enzyme activity:
    250 U
  • Purity:
    ≥98% (SDS-PAGE)
  • Concentration:
    100 nM
  • Formulation:
    In 100 mM TRIS, pH 9,0, containing 200 mM sodium chloride, 1 mM β-mercaptoethanol and 10% glycerol.

Specifications

About this item

Recombinant human matriptase (aa 596-855) fused at the N-terminus to a His-tag.