Human recombinant Caspase-8 (from E. coli)
Produced in E. coli.
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Form:Lyophilised
- Reconstitution instructions:Reconstitute to 1 U/µl with PBS containing 15% glycerol.
- Enzyme name:Caspase-8
- Purity:≥95% (SDS-PAGE)
Frequently Bought Together