Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Caspase-9 (from E. coli)
Human recombinant Caspase-9 (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant Caspase-9 (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Form:
    Lyophilised
  • Reconstitution instructions:
    Reconstitute to 1 U/µl with 25 µl or 100 µl PBS containing 15% glycerol. Note: It is normal to see some insoluble protein following reconstitution. This is mainly due to denaturation of small amounts of protein during preparation. Precipitate should not affect activity. If preferred, remove precipitate by brief centrifugation.
  • Enzyme name:
    Caspase-9
  • Enzyme activity:
    25 U
  • Purity:
    ≥90% (SDS-PAGE)

Specifications

Frequently Bought Together