Human recombinant IFN-alpha-2b (active)
Active enzyme with antiviral and antiproliferative properties.
Produced in HEK cells. The human cytokine (aa 24-188) is fused at the N-terminus to a FLAG®-tag followed by an 8 AA linker.
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:Interferon-alpha-2b|IFNa2b
- Protein/peptide name:IFN-alpha-2b (active)
- Purity:>90% (SDS-PAGE)
- Molecular weight:~21 kDa
- Concentration:0,1 mg/ml when reconstituted as suggested.
- Formulation:Lyophilized. In PBS.
- Purification:Affinity purified.
Frequently Bought Together