Human recombinant IL-1alpha
Catalog # ENZOENZPRT1860010
Fournisseur: ENZO LIFE SCIENCES
Numéro CAS:
Spécifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Biological activity:Stimulates the proliferation of mouse D10S cells.
- Protein synonyms:Hematopoietin-1|Endogenous pyrogen|IL-1F1|LAF|Leukocyte endogenous mediator|LEM|Interleukin-1alpha|MCF|Lymphocyte-activating factor |Mononuclear cell factor
- Protein/peptide name:IL-1alpha
- Purity:>98% (SDS and HPLC)
- Molecular weight:~18,0 kDa
- Formulation:Lyophilized from 10 mM TRIS, pH 8,0, containing 50 mM NaCl.
- Purification:Sterile filtered through a 0,2 micron filter.
Spécifications
À propos de cet article
Produced in E. coli.