Human recombinant CD36
Catalog # ENZOENZPRT1700010
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:
- Species:Human
- Seq:02
- Environmentally Preferable:
- Protein synonyms:GP4|GP3B|SCARB3|Platelet glycoprotein 4
- Protein/peptide name:CD36
- Purity:>95% (SDS-PAGE)
- Molecular weight:~47,5 kDa
- Formulation:Lyophilized from 0,22 µm filtered solution in PBS, pH 7,4. Generally 5 - 8% mannitol or trehalose is added as a protectant before lyophilization.
Specifications
About this item
Produced in HEK 293 cells.