Order Entry
Belgium
ContactUsLinkComponent
Shrimp recombinant alkaline phosphatase
Shrimp recombinant alkaline phosphatase
Catalog # ENZOENZPRT1611000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Shrimp recombinant alkaline phosphatase
Catalog # ENZOENZPRT1611000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Protein/peptide type:
    Recombinant
  • Species:
    Shrimp
  • Seq:
    02
  • Protein synonyms:
    ligation|dNTPs|dephosphorylation|restriction enzyme|cloning|vector|PCR clean-up|SAP|sequencing|plasmid|rSAP
  • Protein/peptide name:
    Alkaline phosphatase
  • Molecular weight:
    54 kDa
  • Concentration:
    1 U/µl for 1000 unit size 5 U/µl for 5000 unit size
  • Formulation:
    Liquid
  • Purification:
    Purified by SDS-PAGE. DNase, RNase or protease activities not detected

Specifications

About this item

Heat-labile alkaline phosphatase for dephosphorylation of DNA, RNA and nucleotides

  • Very high specific activity
  • Completely heat-inactivated at 65°C
  • Removes 5’-phosphates from DNA, RNA, dNTPs and proteins
  • Works in restriction enzyme buffers

Pandalus borealis (Northern red shrimp) origin, recombinantly produced in Pichia pastoris.
Shrimp alkaline phosphatase (recombinant), (rSAP) is a multipurpose alkaline phosphatase that is fully inactivated by a short heat treatment. rSAP has all the properties of the native enzyme, but with the additional benefits of greater stability, consistency, and purity.rSAP simplifies cloning and sequencing workflows due to a complete heat inactivation.