Human recombinant GDNF
Catalog # ENZOENZPRT1450010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein synonyms:Glial cell-derived neurotrophic factor|ATF|Astrocyte-derived trophic factor|hGDNF
- Protein/peptide name:GDNF
- Purity:>98% (SDS-PAGE gel & HPLC)
- Formulation:Lyophilised from 10 mm sodium citrate, containing 150 mm NaCl
Specifications
About this item
Produced in E.coli.