Human recombinant IFN-gamma
Catalog # ENZOENZPRT1410100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein synonyms:Interferon-γ
- Protein/peptide name:IFN-gamma
- Purity:>98% (SDS-PAGE gel & HPLC)
- Molecular weight:~16.8 kDa
- Formulation:Lyophilised from 10 mm sodium phosphate, pH 7,4
Specifications
About this item
Produced in E.coli.