Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant IL-15
Mouse recombinant IL-15
Catalog # ENZOENZPRT1400010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mouse recombinant IL-15
Catalog # ENZOENZPRT1400010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Seq:
    02
  • Protein synonyms:
    Interleukin-15
  • Protein/peptide name:
    IL-15
  • Purity:
    >98% (SDS-PAGE gel & HPLC)
  • Molecular weight:
    ~13.3 kDa
  • Formulation:
    Lyophilized from 10 mm sodium phosphate, pH 8,0

Specifications

About this item

Produced in E.coli.