Human recombinant NEDD8 activating enzyme E1,HIS tag
Catalog # ENZOENZPRT1120050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein synonyms:Ubiquitin-like modifier-activating enzyme 3|Ubiquitin-activating enzyme 3|NEDD8-activating enzyme E1 catalytic subunit|NEDD8-activating enzyme E1C|Ubiquitin-activating enzyme E1C
- Protein/peptide name:NEDD8 activating enzyme E1
- Purity:>95% (SDS-PAGE)
- Molecular weight:62 kDa + 53 kDa (heterodimer: 112,15 kDa)
- Formulation:Liquid, In 25 mM HEPES, pH 7,5, containing 150 mM NaCl, 1 mM DTT
- Purification:Copurified in His-tagged form with Uba3; copurified with APP-BP1
Specifications
About this item
Produced in E. coli.