Order Entry
Belgium
ContactUsLinkComponent
Human recombinant yellow fever NS1
Human recombinant yellow fever NS1
  ENZOENZPRT1090100
 :  
Human recombinant yellow fever NS1
  ENZOENZPRT1090100
 :  

 

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein synonyms:
    non-structural protein|YFV NS1|viral|Strain 17D
  • Protein/peptide name:
    Yellow Fever NS1
  • Purity:
    >95% (SDS-PAGE)
  • Formulation:
    Liquid, In DPBS pH 7,4
  • Purification:
    Purified protein

 

 

Yellow Fever virus NS1 protein presented in its native folded state with all post-translational modifications to deliver optimal antigenicity.

Produced in HEK 293 cells. Yellow Fever Virus NS1 protein from strain 17D.