Recombinant dengue NS1 ST2
Catalog # ENZOENZPRT1050100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Seq:02
- Protein synonyms:non-structural protein|Thailand/16681/84|viral|DENV1-NS1
- Protein/peptide name:Dengue NS1 ST2
- Purity:>95% (SDS-PAGE)
- Formulation:Liquid, In DPBS pH 7,4
- Purification:Purified protein
Specifications
About this item
Dengue virus NS1 protein serotype 2 presented in its native folded state with all post-translational modifications to deliver optimal antigenicity.
Produced in HEK 293 cells. Dengue Viral NS1 Serotype 2 (DENV2-NS1) from strain Thailand/16681/84.